This product is now discontinued, View Related Products
Thermo Scientific™ Hypersil™ BDS Cyano HPLC Columns
Long life and trusted reproducibility for improved analyses
  • pH stable from 2 to 9
  • Base deactivating and endcapping reduce peak tailing for more precise results
Thermo Scientific™ Hypersil™ BDS (Base Deactivated Silica) columns have been trusted worldwide for over 20 years. Each column is manufactured with the same exactness and come with a certificate of analysis, ensuring you that each column will perform as promised. These high standards of manufacturing provide you with consistency and reliability year after year, meaning that you will not have to worry about changing procedures when you need to replace your column.
Hypersil BDS Cyano Columns can be used in either reversed phase or normal phase applications — reversed phase offers different selectivity when compared to C8 or C18 columns and in normal phase these columns show less retention than silica columns.
$686.16 - $1,485.18CAD / Each
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific Hypersil 28803-052130 BDS Cyano Column - Length 50mm, 2.1mm ID, 3µm particle size RK-01886-18 Mfr # 28803-052130
COMPARE
3
$793.42CAD / Each
Thermo Scientific Hypersil 28803-053030 BDS Cyano Column - Length 50mm, 3.0mm ID, 3µm particle size RK-01886-19 Mfr # 28803-053030
COMPARE
3
$793.42CAD / Each
Thermo Scientific Hypersil 28803-054030 BDS Cyano Column - Length 50mm, 4.0mm ID, 3µm particle size RK-01886-20 Mfr # 28803-054030
COMPARE
3
$753.27CAD / Each
Thermo Scientific Hypersil 28803-054630 BDS Cyano Column - Length 50mm, 4.6mm ID, 3µm particle size RK-01886-21 Mfr # 28803-054630
COMPARE
3
$723.43CAD / Each
Thermo Scientific Hypersil 28803-102130 BDS Cyano Column - Length 100mm, 2.1mm ID, 3µm particle size RK-01886-22 Mfr # 28803-102130
COMPARE
3
$860.46CAD / Each
Thermo Scientific Hypersil 28803-103030 BDS Cyano Column - Length 100mm, 3.0mm ID, 3µm particle size RK-01886-23 Mfr # 28803-103030
COMPARE
3
$860.46CAD / Each
Thermo Scientific Hypersil 28803-104030 BDS Cyano Column - Length 100mm, 4.0mm ID, 3µm particle size RK-01886-24 Mfr # 28803-104030
COMPARE
3
$818.47CAD / Each
Thermo Scientific Hypersil 28803-104630 BDS Cyano Column - Length 100mm, 4.6mm ID, 3µm particle size RK-01886-25 Mfr # 28803-104630
COMPARE
3
$786.05CAD / Each
Thermo Scientific Hypersil 28803-152130 BDS Cyano Column - Length 150mm, 2.1mm ID, 3µm particle size RK-01886-26 Mfr # 28803-152130
COMPARE
3
$907.98CAD / Each
Thermo Scientific Hypersil 28803-153030 BDS Cyano Column - Length 150mm, 3.0mm ID, 3µm particle size RK-01886-27 Mfr # 28803-153030
COMPARE
3
$907.98CAD / Each
Thermo Scientific Hypersil 28803-154030 BDS Cyano Column - Length 150mm, 4.0mm ID, 3µm particle size RK-01886-28 Mfr # 28803-154030
COMPARE
3
$864.88CAD / Each
Thermo Scientific Hypersil 28803-154630 BDS Cyano Column - Length 150mm, 4.6mm ID, 3µm particle size RK-01886-29 Mfr # 28803-154630
COMPARE
3
$864.88CAD / Each
Thermo Scientific Hypersil 28805-052130 BDS Cyano Column - Length 50mm, 2.1mm ID, 5µm particle size RK-01886-33 Mfr # 28805-052130
COMPARE
5
$720.34CAD / Each
Thermo Scientific Hypersil 28805-053030 BDS Cyano Column - Length 50mm, 3.0mm ID, 5µm particle size RK-01886-34 Mfr # 28805-053030
COMPARE
5
$720.34CAD / Each
Thermo Scientific Hypersil 28805-054030 BDS Cyano Column - Length 50mm, 4.0mm ID, 5µm particle size RK-01886-35 Mfr # 28805-054030
COMPARE
5
$686.16CAD / Each
Thermo Scientific Hypersil 28805-054630 BDS Cyano Column - Length 50mm, 4.6mm ID, 5µm particle size RK-01886-36 Mfr # 28805-054630
COMPARE
5
$686.16CAD / Each
Thermo Scientific Hypersil 28805-102130 BDS Cyano Column - Length 100mm, 2.1mm ID, 5µm particle size RK-01886-37 Mfr # 28805-102130
COMPARE
5
$783.11CAD / Each
Thermo Scientific Hypersil 28805-103030 BDS Cyano Column - Length 100mm, 3.0mm ID, 5µm particle size RK-01886-38 Mfr # 28805-103030
COMPARE
5
$783.11CAD / Each
Thermo Scientific Hypersil 28805-104030 BDS Cyano Column - Length 100mm, 4.0mm ID, 5µm particle size RK-01886-39 Mfr # 28805-104030
COMPARE
5
$747.38CAD / Each
Thermo Scientific Hypersil 28805-104630 BDS Cyano Column - Length 100mm, 4.6mm ID, 5µm particle size RK-01886-40 Mfr # 28805-104630
COMPARE
5
$717.54CAD / Each
Showing 1 - 20 of 28