Thermo Scientific Heratherm Advanced Protocol Security (APS) Mechanical Convection Ovens
Extra layer of protection when process reliability and security are most important
  • Door lock prevents disruption, tampering, or accidental opening
  • Door alarm notifies when door is accidentally left open
  • Automatic undertemperature alarm ensures samples are kept at the correct temperature
Security mechanical convection ovens offer additional safety features for essential sample protection. Adjustable air circulation ensures accurate, even heating from 50 to 330°C. The adjustable fan can be set at speeds best for the application — highest for best temperature stability and uniformity, lowest for minimal airflow. Programmable controller with large easy-to-view display and touch-button controls can store up to 10 programs with 10 discrete steps. Timer function can be set to run on weekly, hourly, or real time.
Auto-dry function deactivates the oven when samples are dry, saving energy. Additional features include a socket for an independent sensor that when connected is shown on display, an electronically controlled damper, a boost feature for rapid heating of cold oven, and a dedicated access port for the introduction of external temperature sensors.
Smooth stainless steel inner chamber has rounded corners for easy cleaning. The door opens 180°, providing easy access to interior for cleaning and loading of probes. A standard overtemperature alarm and additional undertemperature alarm ensures your samples are always protected. The RS-232 interface lets you connect to your computer for documentation of parameters.
$7,072.27 - $9,753.84CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Thermo Scientific Heratherm 51028127 APS Mechanical Oven; 2.0 cu ft, coated steel, 120V RK-38800-66 Mfr # 51028127
COMPARE
2.2 120
$7,072.27CAD / Each
Thermo Scientific Heratherm 51028326 APS Mechanical Oven; 3.5 cu ft, 304 SS, 208-240V RK-38800-74 Mfr # 51028326
COMPARE
3.5 208 / 240
$9,252.89CAD / Each
Thermo Scientific Heratherm 51028540 APS Mechanical Oven; 6.1 cu ft, 304 SS, 208-240V RK-38800-76 Mfr # 51028540
COMPARE
6.1 208 / 240
$9,753.84CAD / Each