This product is now discontinued, View Related Products
Thermo Scientific HERAcell 150i Carbon Dioxide Incubators
100% pure solid copper interior offers natural antimicrobial protection on contact
  • TC sensor for accurate control where temperature and humidity values are consistent
  • IR sensor for applications where temperature and humidity values fluctuate
These air-jacketed incubators are designed to protect your valuable samples while optimizing cell growth and providing superior recovery rates. A unique 100% pure copper interior and an innovative moist heat decontamination technology combine to eliminate microbial growth and ensure worry-free operation and cleaning. The door-mounted iCAN™ touch-screen control simplifies operation and enables rapid access to important information through simple icons and menu prompts. Graph performance trends, run event history logs, and have visibility to changes in the culture environment all from this simple-to-use system.
High-quality sensors are mounted directly within the chamber for precise environmental measurements. A highly efficient fan-assisted convection ensures the uniformity of critical temperature, humidity, and CO2 no matter where samples are located within the unit. A choice of TC or IR gas sensors allows you to tailor the unit to your preference and experimental needs. A unique directly heated water reservoir provides a high relative humidity and helps with rapid recovery after door openings. A water level sensor with audible and visual alerts lets you know when the level gets low and needs to be refilled.
$18,978.74 - $23,753.99CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Thermo Scientific CO2 Incubator, 5.3 cu ft Cu/TC; 120V RK-39321-00 Mfr # 51026283
COMPARE
120
$18,978.74CAD / Each
Thermo Scientific CO2 Incubator, 5.3 cu ft Cu/IR; 120V RK-39321-02 Mfr # 51026534
COMPARE
120
$23,753.99CAD / Each