This product is now discontinued, View Related Products
Thermo Scientific GENESYS 10S Bio UV/Visible Scanning Spectrophotometer
Versatile, accurate UV/Vis for life science applications
  • Pre-configured methods for nucleic acid and protein analysis
  • No warm up time needed — instant measurements from start up
  • USB connectivity to computers, printers, and data storage devices
  • Use Smart Screen to design a power-up menu that displays frequently-used tests
This spectrophotometer offers the flexibilty and performance a busy laboratory demands. The time-saving software allows you to measure DNA, RNA, and oligo concentrations. With the built-in Oligo calculator, determine molecular weight and theoretical Tm. Measure protein concentrations using Bradford, Lowry, BCA, and Biuret reagents. High-intensity xenon lamp and dual-beam optical geometry provide unsurpassed data quality.
USB connectivity allows easy access to USB memory devices. Data can be printed out using either a PCL printer or the optional internal printer.
$9,392.86 - $10,240.06CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Bandwidth Max Wavelength (nm) Min Wavelength (nm)
Availability Pricing
Thermo Scientific 840-207700 Bio UV/Viible pectrophotometer RK-02654-00 Mfr # 840-207700
COMPARE
1.8 nm 1100 190
$9,392.86CAD / Each
Thermo Scientific 840-207800 Bio UV/Vi pectrophotometer with Printer RK-02654-02 Mfr # 840-207800
COMPARE
1.8 nm 1100 190
$10,240.06CAD / Each