This product is now discontinued, View Related Products
Thermo Scientific Gas Chromatography Autosamplers
Automate your process and increase productivity
  • Handles liquid samples with minimal effort
Available in either an 8-vial auto-injector or 105-vial auto-sampler, both units are compatible with Thermo Scientific FOCUS GC 34010-00 or 34010-10. Units may be set-up via user interface or through a dedicated data system such as ChromQuest.
Automatic features on both units include selection of sample withdrawal speed, injection speed selection, filling speed, vacant vial recognition, and addition of 1µL air gap. Unit includes fixed guides for quick positioning of the auto-sampler, as well as self alignment of needle over injector and vial. Each unit features a safety switch that is activated when front door is opened, ensuring you and your sample remain safe.
Auto-injector 34010-30 includes an 8-position sample tray, set of 10 screw-top vials (2-mL) with septa, set of
$8,354.12 - $13,289.98CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Air Pressure (psig) Max Flow Rate (mL/min) Purity
Availability Pricing
Thermo Scientific 25117551 8-position Gc Auto-injector RK-34010-30 Mfr # 25117551
COMPARE
$8,354.12CAD / Each
Thermo Scientific 25117461 105-position Gc Auto-sampler RK-34010-35 Mfr # 25117461
COMPARE
$13,289.98CAD / Each