This product is now discontinued, View Related Products
Thermo Scientific Compact Digital Waving Shaker
Adjustable tilt angle from 0 to 13° ensures maximum mixing efficiency
  • Compact design easily fits in incubators (non-CO2), chambers, and refrigerators (up to 60°C)
  • PID control for a consistent smooth waving motion
  • Digital speed control easily adjusts from 8 to 100 rpm
  • Use in timer mode up to 99 hr, 59 min or continuous rocking
$1,896.25 - $1,915.41CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Load (lbs) Max Speed (rpm) Power (VAC)
Availability Pricing
Thermo Scientific 88880021 Compact Digital Waving Shaker, 100-240 VAC; US Plug RK-04725-08 Mfr # 88880021
COMPARE
11 100 100 to 240
$1,896.25CAD / Each
Thermo Scientific 88880022 Compact Digital Waving Shaker, 100-240 VAC; Int’nl Plug RK-04725-09 Mfr # 88880022
COMPARE
11 100 100 to 240
$1,915.41CAD / Each