This product is now discontinued, View Related Products
Thermo Scientific Barnstead LabTower TII Water Purification Systems
The Barnstead LabTower TII system delivers up to 240L/day of Type 2 pure water (10-15 megohm) from a tap feed water supply. These units are available in 2 product water output sizes, 20 LPH or 40LPH.
LabTower TII system applications requiring large volumes of type 2 water such as reagent preparation, media preparation and glassware rinsing can be met.
LabTower TII system standard features include dispensing of type 2 water from the integrated 100L reservoir or from the system, quick connect cartridges, universal power supply, variable flow control knob, reservoir level controller, pressure regulator, RS232, and core consumables included with the system. Optional features include choice of systems with or without built in UV lamp, external printer, and IOQ documentation (order each separately).
$21,143.15 - $26,830.43CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity ASTM Water Grade With UV Lamp
Availability Pricing
Thermo Scientific Barnstead 50132191 LabTower TII 20 l/hr with 100 L tank RK-99290-52 Mfr # 50132191
COMPARE
20 L/hr Type II No
$22,955.42CAD / Each
Thermo Scientific Barnstead 50132196 LabTower TII 40 l/hr with 100 L tank RK-99290-54 Mfr # 50132196
COMPARE
40 L/hr Type II Yes
$25,327.57CAD / Each
Thermo Scientific Barnstead 50132193 LabTower TII UV 20 l/hr with 100 L tank RK-99290-62 Mfr # 50132193
COMPARE
20 L/hr Type II No
$21,143.15CAD / Each
Thermo Scientific Barnstead 50132141 LabTower TII UV 40 l/hr with 100 L tank RK-99290-64 Mfr # 50132141
COMPARE
40 L/hr Type II Yes
$26,830.43CAD / Each