This product is now discontinued, View Related Products
Thermo Scientific Barnstead Classic Stills - Cylindrical Storage Tank
  • Store distilled water in a noncorrosive and nonleachable tin-coated tank
Constructed of copper and coated with tin—a corrosion-resistant coating that does not leach contaminants into the stored water. Cylindrical tank includes a removable cover, water level sight
glass, and tin-coated faucet with hose connection; also provides connection for low-water cutoff. For use with portable and stationary stills. Larger sizes available.
$2,737.56 - $7,099.53CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
ASTM Water Grade Max Production (L/hour) With Ultrafilter
Availability Pricing
Thermo Scientific Barnstead B3043 Storage Tank for Barnstead Metal Classic Stills, 10 gallon RK-99276-60 Mfr # B3043
COMPARE
$7,099.53CAD / Each
Thermo Scientific Barnstead H1000 Floorstand for Barnstead Metal Classic Stills Storage Tank RK-99276-68 Mfr # H1000
COMPARE
$2,737.56CAD / Each