Thermo Scientific Barnstead B-Pure Double-Holder System
Modular design - fill with your choice of cartridges
  • Perfect for general pretreatment of laboratory water
B-PURE™ systems are modular wall-mountable systems for point-of-use deionization. Simply add modules as needed when your water requirements change. All systems accept pre- and posttreatment cartridges. Use pretreatment cartridges to filter water before reverse osmosis, deionization, or distillation. Use posttreatment cartridges at the point of use for filtration of ultrapure water. Easily install and remove cartridges; a quarter twist releases canister. A PURA®-Lite resistivity indicator can be mounted directly to the housing to monitor water purity threshold—order separately.
Double-Holder system accepts disposable cartridges (required) sold separately. System includes a digital resistivity meter for accurate monitoring of water purity and a spigot for easy dispensing.
$5,341.04CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Thermo Scientific Barnstead D4521 Double-Holder System, 120 VAC RK-99253-10 Mfr # D4521
COMPARE
120
$5,341.04CAD / Each