Thermo Scientific ARCTIC Premium Refrigerated Circulating Baths
Rugged and corrosion-resistant baths for application temperatures from –50 to 200°C
  • Microprocessor PID controller for fast and precise temperature control
  • Large selection of bath sizes to meet your application requirements
  • Control head can be indexed 90° left or right for easy viewing
ARCTIC baths with Premium Controllers (PC) encompass all of the features of the SC and AC models, plus add on a large, high-resolution, full-color display. The pump speed adjusts incrementally from 40% to 100% for flow and turbulence control and the 10-button keypad surrounds the display for fast and intuitive controller navigation.
Additional features include ten 30-step ramp programs and temperature/program charting
on the display.
$12,560.65CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
Thermo Scientific PC200-A25 7-12L Circulating Bath, -25 to 200C, 115V RK-12129-12 Mfr # 1575258
COMPARE
200 -25 7-12L Circulating Bath, -25 to 200C, 115V
$12,560.65CAD / Each