Supco Refrigerator/Freezer Temperature Alarms
Protect your valuable samples
  • Built-in audible and visual alarms
  • Selectable alarm delay prevents activation from slight or temporary temperature change
Switch-selectable alarms activate on either a temperature rise above set point or drop below set point. Set alarm for either no delay or one hour delay to prevent alarm from activating with slight temperature change (i.e. in freezers with frequent door openings). The temperature sensor wire has a standard two-prong plug that can be spliced and extended up to 200 feet using 22-gauge wire. Alarms feature remote screw connections for external devices or remote alarms.
$212.54 - $214.01CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Power (VAC)
Availability Pricing
COMPARE
80 -10 110
$214.01CAD / Each
Supco Surface-Mount Refrigeration Alarm, -25 to 30°C, 110 VAC RK-10461-10 Mfr # TA2C
COMPARE
86 -13 110
$212.54CAD / Each