This product is now discontinued, View Related Products
Cole-Parmer® HBC-100 Series Dry Block Temperature Calibrators
Fast heat up and cool down
  • Safe and convenient calibration of wide range of thermal sensors
  • Temperature range from 45°C below ambient up to 140°C
  • Digital control provides temperature uniformity and stability
  • Easily manage all calibration functions with RS-232 interface
These lightweight, portable dry block temperature calibrators are ideal for use in the field. They can be used with a wide range of inserts (not included) with different dimension wells to match numerous probe sizes. Safety features include an automatic cooling fan and overtemperature cutout which turns power off to the heaters if a fault develops or the unit exceeds maximum operating temperature. Temperature can be displayed in Celsius or Fahrenheit.
$9,236.68 - $14,210.85CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Power (VAC) Temperature Accuracy
Availability Pricing
COMPARE
797 230 ±0.54°F (±0.3°C)
$14,210.85CAD / Each
COMPARE
1202 230 0.4°C
$10,858.88CAD / Each
COMPARE
1202 120 0.4°C
$10,858.88CAD / Each
COMPARE
284 230 ±0.54°F (±0.3°C)
$11,445.29CAD / Each
COMPARE
797 230 ±0.54°F (±0.3°C)
$9,236.68CAD / Each