Cole-Parmer® WBU-200 Series Liquid Calibration Baths
Compact and accurate water baths for external circulation or temperature calibration
  • Fully insulated baths offer excellent heat retention
  • Temperature stability of ±0.005°C depending on choice of control unit
These compact calibration baths offer excellent stability over the entire temperature range. Use these baths for external circulation to maintain temperatures of samples in viscometers, photometers, refractometers, fermenters and other reaction vessels.
All models offer high pump performance and exceptional thermal stability from -35°C to 250°C. Add a flow cooler to the system to achieve temperatures down to -35°C. The water baths are fully insulated on all sides and base and are fitted with a cooling coil for connection to a cold water supply for use at temperatures around ambient.
An immersion circulator is required for liquid calibration baths to operate—order Cole-Parmer IC-300 series or other compatible immersion circulators separately.
$2,207.14 - $2,868.69CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Liquid Calibration Bath with Cooling Coil; 12 L
$2,868.69CAD / Each
COMPARE
Liquid Calibration Bath with Cooling Coil; 7 L
$2,641.79CAD / Each
COMPARE
Liquid Calibration Bath with Cooling Coil, 5 L
$2,207.14CAD / Each