Cole-Parmer® IC-300 Series Basic Digital Immersion Circulator
Accurate PID temperature control at a competitive price
  • Temperature range of -40°C to 120°C†
  • Excellent temperature stability: ±0.01°C at 40°C
  • 4-digit setting with a bright LED digital temperature display
  • Suitable for most routine laboratory applications
  • User adjustable over-temperature cut-out
  • Low liquid level cut-out as standard
This versatile immersion circulator can be used in several ways. Use as a stand-alone water bath temperature controller, a pump for recirculating chilled liquid from an external chiller, or a pump for providing temperature control to an external device. The basic digital circulator digital LED display, ±0.01°C stability, overtemperature indicator light, pressure pump, and low-liquid level cutoff. Reach below ambient temperatures by using the cooling coil 12122-87 connected to an external chiller.
$2,548.96CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Description
Availability Pricing
COMPARE
120 115 Basic Digital Immersion Circulator; 120 VAC, 50/60Hz
$2,548.96CAD / Each
COMPARE
120 230 Basic Digital Immersion Circulator; 240 VAC, 50/60Hz
$2,548.96CAD / Each