Cole-Parmer® GT-200 Series Gelation Timers
Measure the gelation time of adhesives and resins for quality control
  • Automatically stops when gelation occurs
  • Audible and visual alarms alert you at the gelation point
  • Simple-to-use push buttons with large LED display
  • Stainless steel plungers remove for easy cleaning
The gelation time is the transition from liquid to solid during polymerization. Test requires approximately 100 mL of sample in a container that is >1-1/2" (38 mm) in diameter. Once the measurement is started, the plunger moves up and down. After gelation, the plunger is halted, stopping the timer and setting off the audible and visual alarms. The bright LED digital display shows the time to the nearest minute or tenth of a minute.
Stand and clamp are sold separately.
$4,953.54 - $5,009.53CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Timer Range (minutes) Power (VAC) Resolution
Availability Pricing
COMPARE
999.9 120 0.1 minutes
$4,971.22CAD / Each
COMPARE
999.9 230 0.1 minutes
$5,009.53CAD / Each
COMPARE
9999 120 1 minute
$4,953.54CAD / Each
COMPARE
9999 230 1 minute
$5,008.05CAD / Each