This product is now discontinued, View Related Products
Techne Brewing Microbe 26S Ribosomal RNA Detection Kit for qPCR
Quickly and accurately detect the presence of Dekkera bruxellensis
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Get accurate results faster and easier with qPCR. While D. bruxellensis is an important contributor to flavor development in beer, it can produce undesirable aromas in wine that can cause enormous economic losses. Select a complete kit with mastermix or a kit without mastermix and provide your own.
$1,440.98 - $1,803.43CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Mastermix Included Test Type Description
Availability Pricing
COMPARE
No DNA Dekkera bruxellensis, without mastermix
$1,440.98CAD / Each
COMPARE
Yes DNA Dekkera bruxellensis, with mastermix
$1,803.43CAD / Each