This product is now discontinued, View Related Products
Talboys Orbital Shaker
Low-profile design easily fits into most fume hoods and incubators
  • Variable-speed microprocessor control provides consistent uniform shaking action
This shaker features a maintenance-free, brushless DC motor that provides reliable service and is great for continuous-duty operation. Spill-resistant design channels fluids away from internal components for protection from leaking liquids. A basic adjustment knob features 1 to 10 dial settings. Ramping feature slowly increases speed to desired set point to avoid splashing. Unit can be run in cold rooms, incubators, and CO2 environments from 32 to 104° F (0 to 40°C).
$1,809.32CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Talboys 980173 Standard Orbital Shaker, 40 to 300 rpm, 120 VAC RK-51800-01 Mfr # 980173
COMPARE
120
$1,809.32CAD / Each
Talboys 980174 Standard Orbital Shaker, 40 to 300 rpm, 230 VAC RK-51800-03 Mfr # 980174
COMPARE
230
$1,809.32CAD / Each