This product is now discontinued, View Related Products
Cole-Parmer® PT+ Toploading Balances with USB Connectivity
Durable cast-aluminum housing with precise weighing satisfies any laboratory application
  • Large weighing surface allows placement of a wide variety of samples
  • Weighing modes include parts counting, percent weighing, animal/dynamic weighing and density determination
  • USB and RS-232 interfaces offer connection to a computer or printer for GLP/GMP-compliant outputs
  • Sealed keypad, splashproof design, and removable stainless steel weighing pan
  • Oversized backlit LCD display provides clear visibility of weighing results
  • Digital filters compensate for vibration, drafts or temperature changes
  • Lock down mounting part offers additional security from theft
  • Selectable multi-language display and output
  • Fast stabilization and response time
  • 2-year warranty
Precision toploading balances are built with the most advanced weighing technology available today, offering durability, and easy-to-use operation. These balances feature USB and RS-232 interfaces which offer connection to a computer or printer for GLP/GMP compliant outputs. The sealed keypad, splashproof design, and removable stainless steel weighing pan offer additional safeguards in a messy lab environment. A lock down mounting point offers additional security from theft.
An oversized backlit LCD display provides clear visibility of weighing results. Digital filters compensate for vibration, drafts, or temperature changes.
$937.08 - $1,775.43CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Capacity (g) Power (VAC) Description
Availability Pricing
COMPARE
210 115 / 230 PT-Series Precision Toploading Balance, 210g x 1mg
$1,171.35CAD / Each
COMPARE
410 115 / 230 PT-Series Precision Toploading Balance, 410g x 1mg
$1,383.51CAD / Each
COMPARE
2200 115 / 230 PT-Series Precision Toploading Balance 2200g x 0.01g
$937.08CAD / Each
COMPARE
4200 115 / 230 PT-Series Precision Toploading Balance 4200g x 0.01g
$1,383.51CAD / Each
COMPARE
5000 115 / 230 PT-Series Precision Toploading Balance 5000g x 0.1g
$1,006.33CAD / Each
COMPARE
15000 115 / 230 PT-Series Precision Toploading Balance 15000g x 0.1g
$1,775.43CAD / Each
COMPARE
213 115 / 230 PT-Series Precision Toploading Balance, 210g x 1mginternal Cal
$1,466.02CAD / Each
COMPARE
413 115 / 230 PT-Series Precision Toploading Balance, 410g x 1mginternal Cal
$1,673.77CAD / Each
COMPARE
2200 115 / 230 PT-Series Precision Toploading Balance 2200g x 0.01ginternal Cal
$1,172.82CAD / Each
COMPARE
4200 115 / 230 PT-Series Precision Toploading Balance 4200g x 0.01ginternal Cal
$1,673.77CAD / Each
Showing 1- 10 of 10