Cole-Parmer® MB-200 Series Halogen-Heated Moisture Balances
Fast, simple, and dependable – ideal for basic moisture determination
  • Large, backlit display shows temperature, time, and moisture simultaneously
  • Results provided in grams, % moisture, % solids, % moisture vs. dry weight, and % solid vs. dry weight
  • Displays and prints in four languages: English, German, French, Spanish
  • Allows testing for up to 99 minutes
  • Pan lifter allows safe removal of hot sample pans
  • Security locking slot for Kensington™ type lock and cable prevents theft
Fast response time and easy-to-use functionality make the MB ideal for a wide range of different applications. An automatic test setting function lets you quickly run multiple tests. The auto test feature starts to analyzes the sample when the lid closes and ends when moisture loss stabilizes. Internal memory lets you store a range of test procedures that can be recalled at a touch of a button to make setup and changing from one sample to another. A soft-closing door protects heating element. A single 400W halogen heater delivers controlled temperatures from 50 to 160°C. Overload protection prevents damage to internal weighing components and offers continued performance and long-term usability.
$3,534.66CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Heater Type Power (VAC)
Availability Pricing
COMPARE
40 Halogen 115
$3,534.66CAD / Each
COMPARE
40 Halogen 230
$3,534.66CAD / Each