Sustayn by Spilfyter Recycled Oil-Only Spill Socks
Sorbent material is 100% recycled polypropylene
Go green and support your environmental initiatives with these 100% recycled polypropylene spill socks. Packaging material is also recycled and printing uses environmentally-friendly water-based inks. Oil-Only sorbents absorb all oil-based liquids and repel water. Ideal for use with lubricants, fuels, mineral oils, and vegetable oils.
$166.49 - $232.06CAD / Case of 40
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Absorption Length (in) Width (in)
Availability Pricing
NPS Sustayn by Spilfyter Recycled Oil-Only Blue Sock, 3" x 4', 40/Case RK-86423-19 Mfr # M-34
COMPARE
44 gal. (167 L) 48 3
$232.06CAD / Case of 40
NPS Sustayn by Spilfyter Recycled Oil-Only Blue Sock, 3" x 10', 6/Case RK-86423-20 Mfr # M-37
COMPARE
17 gal. (65 L) 120 3
$166.49CAD / Case of 6