This product is now discontinued, View Related Products
Survivair Blue 1 Half-Facepiece Respirators
Silicone skirt is flexible, lightweight, and durable
  • Speaking diaphragm improves communication for added safety
These lightweight respirators offer a comfortable, secure fit for excellent protection against organic vapors, acid gases, contaminates, and more. Choose from several styles of cartridges and filters (order separately) according to the demands of your work environment.
Approval: NIOSH
Survivair Blue 1™ Half-Mask Respirators feature dual-flange seal for a secure seal and comfortable fit. Rear split strap makes donning easy while retainer clips hold excess strap in place. Large inhalation port lowers breathing resistance.
$62.40 - $67.78CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Size
Availability Pricing
Honeywell B210010 Half-Mask Respirator, Small RK-81662-60 Mfr # B210010
COMPARE
Small
$62.40CAD / Each
Honeywell B220010 Half-Mask Respirator, Medium RK-81662-61 Mfr # B220010
COMPARE
Medium
$67.78CAD / Each
Honeywell B230010 Half-Mask Respirator, Large RK-81662-62 Mfr # B230010
COMPARE
Large
$67.78CAD / Each