Erie Scientific Superfrost® Plus and Colorfrost® Plus Adhesion Microscope Slides

Tissue sections and cells electrostatically adhere to the glass without adhesives – eliminates tissue loss during staining
  • Color-coded for quick and simple identification
  • Marking surfaces ensure easy labeling
  • Slides measure 75 x 25 mm and are 1-mm thick
A permanent positive charge applied to the slide surface allows slides to hold tissue sections without the need for expensive or time-consuming preparatory steps in the laboratory

This treatment virtually eliminates tissue loss during staining and electrostatically attracts frozen tissue sections and cytology preparations, binding them to the slide.
$231.69 - $233.90CAD / Pkg of 144

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Slide Type
Availability Pricing
Erie Scientific Superfrost® Plus Adhesion Microscope Slides, White; 144/Pk RK-48512-00 Mfr # 4951PLUS-001
COMPARE
White Superfrost Plus
$231.69CAD / Pkg of 144
Erie Scientific Colorfrost® Plus Adhesion Microscope Slides, Blue; 144/Pk RK-48512-02 Mfr # 5951PLUS-001
COMPARE
Blue Colorfrost Plus
$233.90CAD / Pkg of 144
Erie Scientific Colorfrost® Plus Adhesion Microscope Slides, Green; 144/Pk RK-48512-04 Mfr # 6951PLUS-001
COMPARE
Green Colorfrost Plus
$233.90CAD / Pkg of 144
Erie Scientific Colorfrost® Plus Adhesion Microscope Slides, Yellow; 144/Pk RK-48512-06 Mfr # 8951PLUS-001
COMPARE
Yellow Colorfrost Plus
$233.53CAD / Pkg of 144
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat