Cole-Parmer® Heating Block Kits for Hot Plates and Stirring Hot Plates
Turn your existing 6" x 6" hot plate into a heating mantle
  • Safest way to heat round-bottom flasks
  • Flexible modular system for a wide range of sizes up to 500 mL
  • Low-well design is perfect for multi-neck flasks
Designed to heat round-bottom flasks, the heating block system with interchangeable inserts can be used with various Cole-Parmer hot plates and hot plate stirrers with 6" x 6" top plate. The heating block system offers two sizes of base plates (small and medium) which can accommodate up to three block inserts each. A third larger base plate features heat-resistant handles and accepts a single block insert for flasks up to 500 mL. Convenient starter kits feature a base plate with a selection of heating block inserts in various sizes. Chemical- and solvent-resistant anodized block inserts provide a safe method for synthesis of single or multiple reactions and offer equal or better heating performance than silicon oil. The low-well provides excellent temperature transfer without compromising the view of the reaction or risk of cracking the glassware. Each base plate features a hole to house a temperature controller probe.
$1,327.52 - $2,525.39CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Heater Block Kit for Hot Plate or Stirring Hot Plate, Large
$1,812.27CAD / Each
COMPARE
Heater Block Kit for Hot Plate or Stirring Hot Plate, Small, 25/50/100 mL
$1,812.27CAD / Each
COMPARE
Heater Block Kit for Hot Plate or Stirring Hot Plate, Medium, 3x250 mL
$1,327.52CAD / Each
COMPARE
Heater Block Kit for Hot Plate or Stirring Hot Plate, Medium, 100/250 mL
$2,525.39CAD / Each