Cole-Parmer® BH-200 Series Dry Block Heaters
Provides precise heating of tubes and microplates
  • Hot warning light indicates when the block temperature is above 50°C
Dry block heaters are designed for precise heating of test tubes, microcentrifuge tubes, cuvettes, and microtiter plates. The large thermal mass of the aluminum blocks provides very accurate and stable temperature control. The “number of blocks accepted” indicates the number of single-width blocks accepted by each heater. A heater that accepts two single-width blocks will also accept one double-width block.
$1,273.01 - $2,902.58CAD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Control Type Number Of Blocks Power (VAC)
Availability Pricing
COMPARE
Analog; 130°C 2 115
$1,412.98CAD / Each
COMPARE
Analog; 130°C 2 230
$1,273.01CAD / Each
COMPARE
Digital; 130°C 2 115
$1,628.10CAD / Each
COMPARE
Digital; 130°C 2 230
$1,628.10CAD / Each
COMPARE
Digital; 130°C 3 115
$1,893.31CAD / Each
COMPARE
Digital; 130°C 3 230
$1,871.21CAD / Each
COMPARE
Digital; 200°C 2 115
$1,679.66CAD / Each
COMPARE
Digital; 200°C 2 230
$1,687.03CAD / Each
COMPARE
Digital; 200°C 3 115
$1,930.14CAD / Each
COMPARE
Digital; 200°C 3 230
$1,959.61CAD / Each
COMPARE
Dual Control Digital; 130°C 2 115
$2,821.54CAD / Each
COMPARE
Dual Control Digital; 130°C 2 230
$2,858.38CAD / Each
COMPARE
Dual Control Digital; 200°C 2 230
$2,902.58CAD / Each
Showing 1- 13 of 13