Cole-Parmer® INC-200 Series Stuart Clear Incubators
Clear acrylic construction offers full visibility of sample
  • Hinged front door provides easy access to working chamber
  • Each side panel features two 10-mm diameter ports for introducing gases or cables
Clear acrylic top and sides make it easy to view your sample from all sides—ideal for plant propagation, simulation of tropical conditions, and corrosion testing. Forced air circulation and electronic temperature control ensure accurately maintained conditions. These incubators do not have a base so they can be placed directly over complete instruments; order base 01110-97 separately.
Select from analog or digital models. A simple calibrated knob lets you set the temperature on the analog model. Digital model with LCD lets you set and view chamber temperature. Both models have a push switch preset at 37°C. Safety feature cuts off heat at 72°C.
$4,420.17 - $5,068.46CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Control Type Power (VAC)
Availability Pricing
COMPARE
2.1 Analog 120
$4,626.44CAD / Each
COMPARE
2.1 Analog 230
$4,420.17CAD / Each
COMPARE
2.1 Digital 120
$5,068.46CAD / Each
COMPARE
2.1 Digital 230
$4,751.68CAD / Each