This product is now discontinued, View Related Products
Stir-Pak General Purpose Laboratory Mixers
High-quality, reliable mixers featuring full-range performance
  • Keyless chuck
  • LED indicator
Our general-purpose mixer is ideal for routine, light-duty mixing of water-like viscosities. Speed ramps to set speed. The brushed DC motor has a compact speed controller integrated by a custom anodized aluminum potentiometer that controls both on/off and variable-speed settings for single direction mixing. Motor has an IP24 rating.
$904.66 - $1,343.73CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Description
Availability Pricing
COMPARE
4000 9.5 General Laboratory Mixer w/o Stand, 115-230VAC
$904.66CAD / Each
COMPARE
4000 9.5 General Laboratory Mixer Complete System
$1,343.73CAD / Each