This product is now discontinued, View Related Products
Stir-Pak Dual-Shaft Mixers
High-quality, reliable mixers featuring full-range performance
  • Use for either high-speed or high-torque mixing
  • Solid-state—gives you precise control of speed
Two motor shafts give you two mixers in one. The direct-drive shaft provides high-speed, low-viscosity mixing; switch to the gear-reduction shaft for your low-speed, high-viscosity mixing needs.
Rugged, permanent-magnet, totally enclosed, nonventilated (TENV) motor has permanentlylubricated ball bearings. Gear-shaft gear ratio is 20:1.
Our solid-state controller maintains speed regulation better than ±2%—critical for materials that change viscosity as mixing progresses. Improved torque limiting feature protects against motor gear failure. Motor has an IP24 rating; controller is totally enclosed and has an IP22 rating for protection against dust and water. Controller complies with UL and CSA 61010 standards. A 6-ft (1.8-m) cord connects the controller and the motor.
$1,398.25 - $2,092.21CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Dual-Shaft Mixer; 115 VAC, 50/60 Hz
$1,398.25CAD / Each
COMPARE
Dual-Shaft Mixer Complete System, 115 VAC
$2,092.21CAD / Each