This product is now discontinued, View Related Products

Steel Hasps

Ideal when procedures require more than one worker on locked-out equipment
  • Red vinyl-coated handles over zinc-plated steel
  • Accommodates up to six padlocks with shackle diameter up to 9/32" (sold separately)
Manufactured or zinc-plated steel, these hasps feature a red vinyl coated handle. accommodates up to six padlocks with shackle diameter up to 9/32" (7.1 mm).
$10.46 - $24.31CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Jaw Opening (in) Description
Availability Pricing
Accuform KDD107 1" Standard-Style Steel Hasp, Red, Accommodates up to 6 Locks RK-40504-01 Mfr # KDD107
COMPARE
1 1\" Standard-Style Steel Hasp, Red, Accommodates up to 6 Locks
$21.36CAD / Each
Accuform KDD109 1-1/2" Standard-Style Steel Hasp, Red, Accommodates up to 6 Locks RK-40504-03 Mfr # KDD109
COMPARE
1 1/2 1-1/2\" Standard-Style Steel Hasp, Red, Accommodates up to 6 Locks
$10.46CAD / Each
Accuform KDD102 1" Interlocking-Style Steel Hasp, Red, Accommodates up to 6 Locks RK-40504-05 Mfr # KDD102
COMPARE
1 1\" Interlocking-Style Steel Hasp, Red, Accommodates up to 6 Locks
$21.36CAD / Each
Accuform KDD103 1-1/2" Interlocking-Style Steel Hasp, Red, Accommodates up to 6 Locks RK-40504-07 Mfr # KDD103
COMPARE
1 1/2 1-1/2\" Interlocking-Style Steel Hasp, Red, Accommodates up to 6 Locks
$24.31CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat