This product is now discontinued, View Related Products
StableTemp Undercounter BOD Incubator
Incubator holds up to 114 BOD bottles
  • Microprocessor-based controller mounted on the door for convenient, easy access
  • Digital LED display clearly shows temperature readings and alarm indicator
  • LED alarm alerts you to over/under temperature conditions
  • Incubator includes a chamber safety thermostat set at 62°C
  • Compressor is protected from accidental overload by a relay
  • Chamber interior features a 2 A outlet
  • Bottom rollers make installation, leveling, and moving simple
$7,028.07 - $8,029.98CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
COMPARE
115
$7,028.07CAD / Each
COMPARE
230
$8,029.98CAD / Each