This product is now discontinued, View Related Products
StableTemp General-Purpose Laboratory Refrigerators
Ideal for the demanding requirements of daily lab applications
  • Quiet refrigeration system keeps the noise level down
  • Available with or without chart recorders — in a wide range of capacities
  • Glass or solid doors (depending on model)
Our complete line of general-purpose refrigerators provides the reliability and security essential for the protection of sensitive lab samples in the 1 to 12°C (34 to 53°F) range. All models deliver the performance, security and quality that you depend on with basic features such as a digital temperature controller with intuitive interface, fluorine-free high-density insulation, and automatic defrost. Standard security features include keyed power switch, high/low temperature alarm, easily accessible contacts for backup monitoring, door ajar alarm (except on sliding glass doors), and a control panel with key lock.
Painted high-quality steel exterior and white epoxy interior with stainless steel floor make these refrigerators durable and easy to clean. Easily move, level, and stabilize with the included locking casters. A 1" rear access port makes it simple to monitor a sample with an external probe.
$7,427.36 - $15,544.26CAD / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Number of Doors Refrigerator Capacity (cu ft) Description
Availability Pricing
COMPARE
1 12 Refrigerator, 12 cu ft, glass door, 115V
$7,427.36CAD / Each
COMPARE
1 12 Refrigerator, 12 cu ft, glass door, chart recorder, 115V
$8,809.40CAD / Each
COMPARE
1 23 Refrigerator, 25 cu ft, glass door, 115V
$7,859.06CAD / Each
COMPARE
1 23 Refrigerator, 25 cu ft, glass door, chart recorder, 115V
$9,241.10CAD / Each
COMPARE
1 23 Refrigerator, 25 cu ft, solid door, 115V
$7,859.06CAD / Each
COMPARE
1 23 Refrigerator, 25 cu ft, solid door, chart recorder, 115V
$9,241.10CAD / Each
COMPARE
1 27 Refrigerator, 30 cu ft, glass door, 115V
$8,632.59CAD / Each
COMPARE
1 27 Refrigerator, 30 cu ft, glass door, chart recorder, 115V
$10,016.11CAD / Each
COMPARE
1 27 Refrigerator, 30 cu ft, solid door, 115V
$8,632.59CAD / Each
COMPARE
1 27 Refrigerator, 30 cu ft, solid door, chart recorder, 115V
$10,016.11CAD / Each
COMPARE
2 38 Refrigerator, 38 cu ft, glass door, 115V
$9,843.72CAD / Each
COMPARE
2 38 Refrigerator, 38 cu ft, glass door, chart recorder, 115V
$11,224.29CAD / Each
COMPARE
2 45 Refrigerator, 45 cu ft, glass door, 115V
$10,708.60CAD / Each
COMPARE
2 45 Refrigerator, 45 cu ft, glass door, chart recorder, 115V
$12,087.69CAD / Each
COMPARE
2 49 Refrigerator, 49 cu ft, glass door, 115V
$11,224.29CAD / Each
COMPARE
2 49 Refrigerator, 49 cu ft, glass door, chart recorder, 115V
$12,609.27CAD / Each
COMPARE
2 49 Refrigerator, 49 cu ft, solid door, 115V
$11,224.29CAD / Each
COMPARE
2 49 Refrigerator, 49 cu ft, solid door, chart recorder, 115V
$12,609.27CAD / Each
COMPARE
3 72 Refrigerator, 72 cu ft, glass door, 115V
$14,157.80CAD / Each
COMPARE
3 72 Refrigerator, 72 cu ft, glass door, chart recorder, 115V
$15,544.26CAD / Each
Showing 1- 20 of 20