This product is now discontinued, View Related Products
Spilfyter Standard Oil-Only Sorbent Pillows
Pillows feature large surface area with high capacity absorbency
Catch slow leaks and drips with sorbent pillows. Standard pillows feature above average absorption, strength, and low-linting. Oil-Only sorbents absorb all oil-based liquids and repel water. Ideal for use with lubricants, fuels, mineral oils, and vegetable oils.
$123.47 - $230.59CAD / Case of 20
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Absorption Length (in) Width (in)
Availability Pricing
NPS Spilfyter Standard Oil-Only Spill Pillow, 10" x 10", 20/Case RK-86424-36 Mfr # M-61
COMPARE
10 gal. (38 L) 10 10
$230.59CAD / Case of 20
NPS Spilfyter Standard Oil-Only Spill Pillow, 18" x 18", 10/Case RK-86424-37 Mfr # M-62
COMPARE
13 gal. (50 L) 18 18
$164.65CAD / Case of 10
NPS Spilfyter Standard Oil-Only Spill Pillow, 8.5" x 17", 10/Case RK-86424-38 Mfr # M-66
COMPARE
5 gal. (19 L) 17 8 1/2
$123.47CAD / Case of 10