Cole-Parmer® VapLock™ Solvent Waste Manifold Caps for Drums
Connect solvent waste manifold caps to waste containers for a secure closed system
  • Choose from polypropylene or PTFE construction
Select from various manifold cap sizes to fit your laboratory and industrial solvent waste containers. Drum caps fit on 2" bung opening (fine or coarse thread) and come with PP drum thread adapter. Caps are available with polypropylene (PP) or PTFE manifolds and 4 or 10 variously sized ports. Ports accept male thread adapters to connect flexible and rigid tubing, or filter 12019-79 or filter/indicator 12019-80.
$269.63 - $574.99CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap / Stopper Size Material Special Features
Availability Pricing
COMPARE
2\" Fine Thread Polypropylene(PP) 1x1/2\"NPT, 3x1/4\"NPT, 6x1/4\"-28UNF
$290.63CAD / Each
COMPARE
2\" Fine Thread Polypropylene(PP) 1x1/2\" NPT, 3x1/4\" NPT
$269.63CAD / Each
COMPARE
2\" Fine Thread Polytetrafluoroethylene (PTFE) 1x1/2\"NPT, 3x1/4\"NPT, 6x1/4-28UNF
$574.99CAD / Each
COMPARE
2\" Fine Thread Polytetrafluoroethylene (PTFE) 1x1/2\" NPT, 3x1/4\" NPT
$501.32CAD / Each
COMPARE
2\" Coarse Thread Polypropylene(PP) 1x1/2\"NPT, 3x1/4\"NPT, 6x1/4\"-28UNF
$291.36CAD / Each
COMPARE
2\" Coarse Thread Polypropylene(PP) 1x1/2\" NPT, 3x1/4\" NPT
$269.63CAD / Each
COMPARE
2\" Coarse Thread Polytetrafluoroethylene (PTFE) 1x1/2\"NPT, 3x1/4\"NPT, 6x1/4-28UNF
$468.91CAD / Each
COMPARE
2\" Coarse Thread Polytetrafluoroethylene (PTFE) 1x1/2\" NPT, 3x1/4\" NPT
$478.12CAD / Each
Showing 1- 8 of 8