Cole-Parmer® VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys
Connect solvent waste manifold caps to waste containers for a secure closed system
  • Choose from polypropylene or PTFE construction
Select from various manifold cap sizes to fit your laboratory and industrial solvent waste containers. Bottle and carboy caps are available with polypropylene (PP) or PTFE manifolds and 4 or 10 variously sized ports. Ports accept male thread adapters to connect flexible and rigid tubing, or filter 12019-79 or filter/indicator 12019-80.
$234.64 - $572.78CAD / Each
49 variations of this product are available.
See All Product Options Below
49 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap / Stopper Size Material Special Features
Availability Pricing
COMPARE
GL38 Polypropylene(PP) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F), 6 x 1/4-28 UNF(F)
$292.10CAD / Each
COMPARE
GL38 Polypropylene(PP) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F)
$260.42CAD / Each
COMPARE
GL38 Polytetrafluoroethylene (PTFE) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F), 6 x 1/4-28 UNF(F)
$343.67CAD / Each
COMPARE
GL38 Polytetrafluoroethylene (PTFE) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F)
$328.57CAD / Each
COMPARE
38-430 Polypropylene(PP) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F), 6 x 1/4-28 UNF(F)
$322.30CAD / Each
COMPARE
38-430 Polypropylene(PP) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F)
$236.11CAD / Each
COMPARE
38-430 Polytetrafluoroethylene (PTFE) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F), 6 x 1/4-28 UNF(F)
$390.08CAD / Each
COMPARE
38-430 Polytetrafluoroethylene (PTFE) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F)
$374.61CAD / Each
COMPARE
38 mm Wako Polypropylene(PP) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F), 6 x 1/4-28 UNF(F)
$275.89CAD / Each
COMPARE
38 mm Wako Polypropylene(PP) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F)
$252.32CAD / Each
COMPARE
38 mm Wako Polytetrafluoroethylene (PTFE) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F), 6 x 1/4-28 UNF(F)
$499.85CAD / Each
COMPARE
38 mm Wako Polytetrafluoroethylene (PTFE) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F)
$345.14CAD / Each
COMPARE
GL45 Polypropylene(PP) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F), 6 x 1/4-28 UNF(F)
$282.52CAD / Each
COMPARE
GL45 Polypropylene(PP) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F)
$258.21CAD / Each
COMPARE
GL45 Polytetrafluoroethylene (PTFE) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F), 6 x 1/4-28 UNF(F)
$537.42CAD / Each
COMPARE
GL45 Polytetrafluoroethylene (PTFE) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F)
$393.03CAD / Each
COMPARE
DIN 45 Polypropylene(PP) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F), 6 x 1/4-28 UNF(F)
$297.62CAD / Each
COMPARE
DIN 45 Polypropylene(PP) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F)
$234.64CAD / Each
COMPARE
DIN 45 Polytetrafluoroethylene (PTFE) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F)
$436.86CAD / Each
COMPARE
63 mm Polypropylene(PP) 1 x 1/2\" NPT(F), 3 x 1/4\" NPT(F)
$261.53CAD / Each
Showing 1 - 20 of 49