Cole-Parmer® VapLock™ Solvent Delivery Caps with Inlet Valve and Filter
Reduce chemical evaporation and exposure to hazardous vapors
  • Use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) OD
Create a closed system using solvent reservoir caps to prevent hazardous vapors from escaping into the lab. Caps accept standard 1/4"-28 UNF(M) fittings to easily connect semi-rigid tubing. Anti-twist design allows the inner body of the cap to rotate independently preventing tubes from tangling when caps are removed or attached. Caps are constructed of PTFE body and PP collar* and are autoclavable.
Caps with integrated air inlet valve and filter permit air to enter the bottle for vacuum displacement while solvent is pumped to the instrument. Valves are made of PTFE and 300-series SS components and feature a 50 µm PTFE filter to reduce particulate contamination. Select from FFKM, EPDM, or Viton® valve seal options.
$57.17 - $107.26CAD / Each
23 variations of this product are available.
See All Product Options Below
23 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Material Cap / Stopper Size Special Features
Availability Pricing
COMPARE
EPDM air inlet valve GL45 2 x 1/4\"-28 UNF(F)
$66.30CAD / Each
COMPARE
EPDM air inlet valve GL45 4 x 1/4\"-28 UNF(F)
$64.24CAD / Each
COMPARE
FFKM air inlet valve GL45 2 x 1/4\"-28 UNF(F)
$85.31CAD / Each
COMPARE
FFKM air inlet valve GL45 4 x 1/4\"-28 UNF(F)
$92.68CAD / Each
COMPARE
Viton® air inlet valve GL45 2 x 1/4\"-28 UNF(F)
$57.31CAD / Each
COMPARE
Viton® air inlet valve GL45 4 x 1/4\"-28 UNF(F)
$74.41CAD / Each
COMPARE
FFKM air inlet valve GL38 2 x 1/4\"-28 UNF(F)
$85.16CAD / Each
COMPARE
EPDM air inlet valve GL38 2 x 1/4\"-28 UNF(F)
$57.31CAD / Each
COMPARE
Viton® air inlet valve GL38 2 x 1/4\"-28 UNF(F)
$57.31CAD / Each
COMPARE
FFKM air inlet valve GL38 4 x 1/4\"-28 UNF(F)
$107.26CAD / Each
COMPARE
EPDM air inlet valve GL38 4 x 1/4\"-28 UNF(F)
$64.24CAD / Each
COMPARE
Viton® air inlet valve GL38 4 x 1/4\"-28 UNF(F)
$66.16CAD / Each
COMPARE
EPDM air inlet valve Nalgene 38-430 2 x 1/4\"-28 UNF(F)
$57.17CAD / Each
COMPARE
EPDM air inlet valve Nalgene 38-430 4 x 1/4\"-28 UNF(F)
$62.32CAD / Each
COMPARE
FFKM air inlet valve Nalgene 38-430 2 x 1/4\"-28 UNF(F)
$85.31CAD / Each
COMPARE
FFKM air inlet valve Nalgene 38-430 4 x 1/4\"-28 UNF(F)
$90.17CAD / Each
COMPARE
Viton® air inlet valve Nalgene 38-430 2 x 1/4\"-28 UNF(F)
$65.57CAD / Each
COMPARE
Viton® air inlet valve Nalgene 38-430 4 x 1/4\"-28 UNF(F)
$64.39CAD / Each
COMPARE
EPDM® air inlet valve Wako 38 mm 2 x 1/4\"-28 UNF(F)
$64.53CAD / Each
COMPARE
FFKM® air inlet valve Wako 38 mm 2 x 1/4\"-28 UNF(F)
$80.01CAD / Each
Showing 1 - 20 of 23