Spex® VapLock™ Solenoid-Operated Micro Pumps
Provides a precise, repeatable volume of liquid every time
  • Nonmetallic inert fluid path—ideal for high-purity or aggressive fluids
  • High cycle life—up to 20 million dispense cycles
  • Self-priming up to 51” (dry) and 138” (wet)
  • Rated for continuous-duty operation
With the ability to dispense highly accurate and consistent volume, these micro pumps are ideal for chemical dosing, waste effluent removal, and sterilizing applications. Solenoid-operated pumps move the diaphragm up and down as the solenoid is energized and de-energized to pull liquid through the inlet check valves then expel through the outlet check valve. A diaphragm separates the flow path from the operating mechanism ensuring an inert fluid path. These pumps are suitable use for up to 3000 hours at a 2 Hz cycle rate. Simply connect PTFE-coated lead wires with stripped ends for 12 or 24 VDC operation.
$540.37 - $1,338.57CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Wetted Materials Description
Availability Pricing
COMPARE
PEEK/PTFE/FKM Self-Priming Micro Pump, 20 µL, PEEK/PTFE/FKM; 12 VDC
$1,338.57CAD / Each
COMPARE
PEEK/PTFE/FFKM Self-Priming Micro Pump, 20 µL, PEEK/PTFE/FFKM; 12 VDC
$898.03CAD / Each
COMPARE
PEEK/EPDM/EPDM Self-Priming Micro Pump, 50 µL, PEEK/EPDM/EPDM; 12 VDC
$772.79CAD / Each
COMPARE
PPS/EPDM/EPDM Self-Priming Micro Pump, 60 µL, PPS/EPDM/EPDM; 12 VDC
$870.04CAD / Each
COMPARE
PEEK/EPDM/EPDM Self-Priming Micro Pump, 20 µL, PEEK/EPDM/EPDM; 24 VDC
$870.04CAD / Each
COMPARE
PEEK/EPDM/EPDM Self-Priming Micro Pump, 50 µL, PEEK/EPDM/EPDM; 24 VDC
$870.04CAD / Each
COMPARE
PEEK/PTFE/FKM Self-Priming Micro Pump, 50 µL, PEEK/PTFE/FKM; 24 VDC
$870.04CAD / Each
COMPARE
POM/EPDM/EPDM Self-Priming Micro Pump, 20 µL, POM/EPDM/EPDM; 12 VDC
$540.37CAD / Each
COMPARE
POM/PTFE/FKM Self-Priming Micro Pump, 20 µL, POM/PTFE/FKM; 24 VDC
$601.88CAD / Each
COMPARE
POM/PTFE/FKM Self-Priming Micro Pump, 50 µL, POM/PTFE/FKM; 24 VDC
$621.03CAD / Each
Showing 1- 10 of 10