Cole-Parmer® VapLock™ Breakthrough Color Indicator for Exhaust Filters
Use with exhaust filters to detect end-of-service life
  • Compatible with Cole-Parmer® VapLock™ activated carbon exhaust filters
Color change occurs when the indicator is exposed to organic solvent vapors. Indicator housing is made of acrylic.
$76.17 - $360.24CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
COMPARE
VapLock™ activated carbon exhaust filters Breakthrough Indicator for Exhaust Filters; 1/ea
$76.17CAD / Each
COMPARE
VapLock™ activated carbon exhaust filters Breakthrough Indicator for Exhaust Filters; 5/pk
$360.24CAD / Pkg of 5