This product is now discontinued, View Related Products
Sperian ONE-Fit N95 Particulate Respirators
Molded cup contours with the natural shape of the face, minimizing pressure points
  • Dual headstraps and soft inner lining further increase comfort for long term use
Universal one-size respirators are designed to fit most face shapes and sizes. Durable outer shell prevents the mask from collapse when working in high-humidity environments. Molded-nose bridge ensures a comfortable fit and works well with glasses. Electrostatic polypropylene/polyester fibers provide excellent filter performance in applications such as sanding wood, dust exposure, grinding, metal cutting, and healthcare.
Approval: NIOSH
$29.17 - $29.84CAD / Box of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Rating Description
Availability Pricing
Honeywell 14110444 N95 Particulate Respirators, 20/box RK-81660-40 Mfr # 14110444
COMPARE
N95 N95 Particulate Respirators, 20/box
$29.17CAD / Box of 1
Honeywell 14110445 N95 Particulate Respirators with Exhalation Valve, 10/box RK-81660-41 Mfr # 14110445
COMPARE
N95 N95 Particulate Respirators with Exhalation Valve, 10/box
$29.84CAD / Box of 1