This product is now discontinued, View Related Products
Silver Membrane Filters
Chemically resistant with exceptionally high flow rates
  • Ultrathin membranes measure only 50µm (0.002”) in thickness
  • Excellent chemical and thermal properties
  • Intrinsically bacteriostatic due to the nature of silver
Silver metal membranes have the chemical inertness of pure silver. They resist alcohol, fuels, all organic solvents, and other hydrocarbons including halogenated hydrocarbons. Membranes can be repeatedly autoclaved, steam, or hot air sterilized without losing their effectiveness. Adsorption and absorption is virtually nonexistent due to the chemical and biological inertness of pure silver, and the smooth regular shape of the silver media.
Silver membranes are fiber-free and feature virtually no media migration because they are strong, uniform, porous monolithic structures consisting of metallic crystals. The high porosity (60% open area) provides exceptionally high flow rates. The intrinsic properties of silver create an antimicrobial effect that inhibits the growth of bacteria and other microorganisms on the surface of the membrane. Typical applications include X-ray diffraction scanning electron microscopy, high temperature venting, clarification of liquids, HPLC media preparation, high-purity fluids, and bacterial sampling.
$626.19 - $1,060.84CAD / Pkg of 100
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem specifications
Diameter (mm) Material Pore Size (µm)
Availability Pricing
COMPARE
13 Silver 0.2
$1,052.00CAD / Pkg of 100
COMPARE
25 Silver 0.2
$667.45CAD / Pkg of 50
COMPARE
47 Silver 0.2
$783.84CAD / Pkg of 25
COMPARE
13 Silver 0.45
$1,052.00CAD / Each
COMPARE
25 Silver 0.45
$685.13CAD / Pkg of 50
COMPARE
47 Silver 0.45
$807.42CAD / Pkg of 25
COMPARE
13 Silver 0.8
$1,060.84CAD / Pkg of 100
COMPARE
25 Silver 0.8
$626.19CAD / Pkg of 50
COMPARE
47 Silver 0.8
$783.84CAD / Pkg of 25
COMPARE
13 Silver 1.2
$1,034.32CAD / Pkg of 100
COMPARE
25 Silver 1.2
$632.08CAD / Pkg of 50
COMPARE
47 Silver 1.2
$783.84CAD / Pkg of 25
COMPARE
13 Silver 5
$1,034.32CAD / Pkg of 100
COMPARE
25 Silver 5
$651.24CAD / Pkg of 50
COMPARE
47 Silver 5
$813.31CAD / Pkg of 25
Showing 1- 15 of 15