BriskHeat® Silicone-Rubber Heater Blankets Metal Surfaces with Control-240 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 425°F (218°C)
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications.
Blanket with Control offers extra convenience and functionality with its included adjustable thermostat control. Models for metal surfaces can be adjusted between 50 to 425°F. No additional control is necessary
The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Heaters for metal surfaces have faster heat-up rates due to the 2.5 watts /in2 power density.
$288.78 - $1,044.63CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRL06122ADJB Silicone Heating Blanket with Controller, for Metal, 6x12" , 240 V, 180 W RK-36002-34 Mfr # SRL06122ADJB
COMPARE
12 425 240
$288.78CAD / Each
BriskHeat SRL06242ADJB Silicone Heating Blanket with Controller, for Metal, 6x24" , 240 V, 360 W RK-36002-36 Mfr # SRL06242ADJB
COMPARE
24 425 240
$365.40CAD / Each
BriskHeat SRL06362ADJB Silicone Heating Blanket with Controller, for Metal, 6" x 36" , 240 V, 540 W RK-36002-38 Mfr # SRL06362ADJB
COMPARE
36 425 240
$461.17CAD / Each
BriskHeat SRL12122ADJB Silicone Heating Blanket with Controller, for Metal, 12x12" , 240 V, 360 W RK-36002-40 Mfr # SRL12122ADJB
COMPARE
12 425 240
$360.98CAD / Each
BriskHeat SRL12242ADJB Silicone Heating Blanket with Controller, for Metal, 12x24" , 240 V, 720 W RK-36002-42 Mfr # SRL12242ADJB
COMPARE
24 425 240
$505.37CAD / Each
BriskHeat SRL12362ADJB Silicone Heating Blanket with Controller, for Metal, 12x36" , 240 V, 1080 W RK-36002-44 Mfr # SRL12362ADJB
COMPARE
36 425 240
$751.43CAD / Each
BriskHeat SRL18182ADJB Silicone Heating Blanket with Controller, for Metal, 18x18" , 240 V, 810 W RK-36002-46 Mfr # SRL18182ADJB
COMPARE
18 425 240
$611.46CAD / Each
BriskHeat SRL18362ADJB Silicone Heating Blanket with Controller, for Metal, 18x36" , 240 V, 1472 W RK-36002-48 Mfr # SRL18362ADJB
COMPARE
36 425 240
$1,044.63CAD / Each
Showing 1- 8 of 8