BriskHeat® Silicone-Rubber Heater Blankets Metal Surfaces with Adhesive-240 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 450°F (232°C)
  • Easy-to-apply industrial strength pressure sensitive adhesive backing
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications. The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Heaters for metal surfaces have faster heat-up rates due to the 2.5 watts /in2 power density.
$200.38 - $1,096.20CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRL06122P Silicone Heating Blanket with Adhesive, for Metal, 6x12" , 240 V, 180 W RK-36001-02 Mfr # SRL06122P
COMPARE
12 450 240
$200.38CAD / Each
BriskHeat SRL12122P Silicone Heating Blanket with Adhesive, for Metal, 12x12" , 240 V, 360 W RK-36001-09 Mfr # SRL12122P
COMPARE
12 450 240
$246.06CAD / Each
BriskHeat SRL12242P Silicone Heating Blanket with Adhesive, for Metal, 12x24" , 240 V, 720 W RK-36001-12 Mfr # SRL12242P
COMPARE
24 450 240
$428.76CAD / Each
BriskHeat SRL18182P Silicone Heating Blanket with Adhesive, for Metal, 18x18" , 240 V, 810 W RK-36001-17 Mfr # SRL18182P
COMPARE
18 450 240
$542.21CAD / Each
BriskHeat SRL24242P Silicone Heating Blanket with Adhesive, for Metal, 24x24" , 240 V, 1440 W RK-36001-22 Mfr # SRL24242P
COMPARE
24 450 240
$825.10CAD / Each
BriskHeat SRL24362P Silicone Heating Blanket with Adhesive, for Metal, 24x36" , 240 V, 2160 W RK-36001-24 Mfr # SRL24362P
COMPARE
36 450 240
$1,096.20CAD / Each