Sherwood Scientific Sealed Tub Assemblies
Reduce sample loss
  • Silicon O-ring provides tight seal
Sealed tubs reduce sample loss - ideal for drying smaller particles. To avoid overflow of sample into the filter bag, the filter plate seals on a silicon O-ring, retaining the sample in the tub.
$5,753.59 - $7,285.91CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Sherwood Scientific 50135020 Sealed Tub Assembly, with port for temperature/humidity probe; 5-liter RK-02658-01 Mfr # 50135020
COMPARE
Sealed Tub Assembly, with port for temperature/humidity probe; 5-liter
$7,285.91CAD / Each
Sherwood Scientific 50035014 Sealed Tub Assembly, without Port, Tub and Base; 5 Liter, Glass RK-02658-64 Mfr # 50035014
COMPARE
Sealed Tub Assembly, without Port, Tub and Base; 5 Liter, Glass
$5,753.59CAD / Each