SciMatCo Mini Stak-a-Cab Storage Cabinets
Compact size is perfect for laboratory benchtop chemical storage
Rugged cabinets are constructed of 1” (2.5 cm) thick, high-density plywood and finished with clear epoxy paint for superior durability. Liquid-tight, 2” (5.1 cm) trough at base of cabinet contains any spillage. Color and highly visible 8” (20.3 cm) hazard label ensures easy identification of cabinet type. These versatile cabinets are designed for the laboratory bench but also can be stacked three high, using optional floor stand, for a complete chemical storage center.
$1,603.05CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Description
Availability Pricing
SciMatCo SC9042 Storage Cabinet, Acid, Mini 20 L RK-86434-00 Mfr # SC9042
COMPARE
5.3 Storage Cabinet, Acid, Mini
$1,603.05CAD / Each