This product is now discontinued, View Related Products
Scientific RTD Thermometer
A highly accurate thermometer that is simple to use
  • NIST-traceable certificate
This high-precision thermometer is ideal for measuring temperatures in gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, foods and wastewater. Designed for years of reliable service even in severe environments, the meter features a splash proof membrane keypad. A high-contrast 1/2"-high LCD makes the thermometer easy to read in all types of light. The meter features memory and hold function, internal datalogger that captures the min/max temperature every hour for a 24-hours period, and °F/°C selection. For easy reading, the meter can be changed to display one, two, or three significant numbers after the decimal point. The fast-response general purpose probe (included) displays even the slightest temperature change instantly. Readings are updated three times a second. The internal memory can be downloaded into a computer using optional software 90080-21.
$612.93CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Meter Measurement Type Number Of Probe Channels
Availability Pricing
COMPARE
932 RTD 1
$612.93CAD / Each
COMPARE
392 RTD 1
$612.93CAD / Each