Scientific Industries Mini-100 Orbital-Genie™ Variable Speed Shakers
Gently swirl delicate samples
  • Low speed shaking mixes without damaging cells or other fragile sample components
  • Nonslip platform with spill-proof lip keeps vessels secure and your workspace clean
  • Stack up to two optional trays to increase throughput
The Mini-100 Orbital-Genie shaker features a small orbit and low speed to avoid over-agitating or disrupting your samples. Analog speed control allows you to optimize mixing for your application. Included thermoplastic platform has a non-slip rubber mat and raised molded edges to contain spills.
Optional stacking trays double or triple the mixing capacity.
$1,688.50 - $1,697.35CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Description
Availability Pricing
Scientific Industries Mini-100 Orbital-Genie Variable Speed Shaker; 120 VAC, US plug RK-51820-01 Mfr # SI-M100
COMPARE
120 Orbital-Genie Variable Speed Shaker; 120 VAC, US plug
$1,688.50CAD / Each
Scientific Industries Mini-100 Orbital-Genie Variable Speed Shaker; 230 VAC, EU plug RK-51820-03 Mfr # SI-M102
COMPARE
230 Orbital-Genie Variable Speed Shaker; 230 VAC, EU plug
$1,697.35CAD / Each