ScienTemp Compact Ultra-Low Freezers
An ultra-low freezer that can sit on the benchtop!
  • Key locking door provides additional sample security
These compact ultra-low freezers are ideal for research applications and small sample storage. They fit easily on the benchtop or undercounter without taking up a lot of space. Freezers have a temperature range of –50 to –86°C and provide 1.0 cubic feet of storage space. Microprocessor control provides temperature accuracy of ±1°C and features an audible alarm and diagnostic capabilities.
Cabinets include CFC-free, foamed-in-place insulation that conserves energy and protects samples. Both benchtop and undercounter models have the display located at the top of the unit for easy viewing along with a top-mounted compressor. Chart recorders may be ordered separately. Freezers will accommodate up to four storage racks.
$9,577.04CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Freezer Capacity (cu ft) Power (VAC)
Availability Pricing
ScienTemp 86-01A Compact ultra-low freezer, benchtop, 115 VAC, 60 Hz RK-44204-00 Mfr # 86-01A
COMPARE
1 120
$9,577.04CAD / Each