Scienceware SPINFIN STIRRING BAR PTFE
Vigorously shakes liquids in round-bottom test tubes and flasks, where other stir bars cannot be used. Position is maintained in a vessel and will not vibrate.
$16.65 - $19.60CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Description
Availability Pricing
Scienceware F371250012 Spinfin Stirring Bar, PTFE, 1/2" X 1/2" RK-04775-27 Mfr # F371250012
COMPARE
1.9685 Spinfin Stirring Bar, PTFE, 1/2\" X 1/2\"
$19.60CAD / Each
Scienceware F371250014 Spinfin Stirring Bar, PTFE, 5/16" X 3/8" RK-04775-29 Mfr # F371250014
COMPARE
0.375 Spinfin Stirring Bar, PTFE, 5/16\" X 3/8\"
$16.65CAD / Each
Scienceware F371250038 Spinfin Stirring Bar, PTFE, 3/8" X 3/8" RK-04775-33 Mfr # F371250038
COMPARE
0.375 Spinfin Stirring Bar, PTFE, 3/8\" X 3/8\"
$18.12CAD / Each