This product is now discontinued, View Related Products
Scanning IR Spectrophotometer
Touch screen display lets you view spectra and run survey scans without a PC Display measurement values as ratios in either percent transmission or absorbance Single beam optical design provides dual beam capabilities Nitrogen purge not required—eliminate costly gases Connects easily to a computer or recorder
Simply stated, this is the easiest IR spectrophotometer to operate and maintain! Advanced self-help menus guide you every step of the way. Analysis requires no expensive nitrogen purge for moisture or CO2 prior to use. Single-button controls set all instrument parameters for operation and rapid data collection. High energy optical design and a sensitive DTGS detector provides excellent resolution over the full infrared range. Maximum performance is achieved with a fast scanning mechanism that is completely microprocessor driven—you get dual-beam capabilities with just a single beam optical bed. Display simultaneously indicates wavelength, transmittance or absorbance, scan type, scan time, and response time in seconds. Easy Scan software (included) easily accomm
$4,641.18 - $17,525.97CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Bandwidth Source Lamp Wavelength Accuracy
Availability Pricing
Buck Scientific BUCK M500 Scanning Infrared Spectrophtometer; 115/230V RK-83000-00 Mfr # M530
COMPARE
4000 to 600 cm-1 Nichrome wire-wound ceramic core ±3 cm-1
$17,525.97CAD / Each
Buck Scientific GRAMS/AI (version 7.0) Infrared Analysis Software for Windows RK-83001-27 Mfr # GRAMS/AI
COMPARE
$4,641.18CAD / Each