Sample Vials with Threaded Closures, PFA
Sample vials are ideal for sensitive samples
  • Constructed of chemically inert PFA
PFA vials are inert and nonleaching, with a temperature range of –320 to 500°F (–195 to 260°C)—ideal for sensitive samples. Choose from vials with or without organizing tray. Trays are black HDPE with numbered pockets and slots for identification cards (not included). Each tray contains ten vials.
$35.80 - $1,085.89CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Type Thread Volume (mL)
Availability Pricing
Sample Vial, PFA, without Organizing Tray or Cap, 18 mm Thread, 4 mL RK-06103-06 Mfr # 200-004-13
COMPARE
No cap included 18 mm 4
$58.20CAD / Each
Sample Vial, PFA, without Organizing Tray or Cap, 24 mm Thread, 7 mL RK-06103-08 Mfr # 200-007-10
COMPARE
No cap included 24 mm 7
$35.80CAD / Each
Sample Vial, PFA, without Organizing Tray or Cap, 33 mm Thread, 15 mL RK-06103-09 Mfr # 200-015-12
COMPARE
No cap included 33 mm 15
$62.77CAD / Each
Sample Vial, PFA, without Organizing Tray or Cap, 33 mm Thread, 22 mL RK-06103-11 Mfr # 200-022-12
COMPARE
No cap included 33 mm 22
$74.85CAD / Each
Sample Vial, PFA, without Organizing Tray or Cap, 33 mm Thread, 30 mL RK-06103-12 Mfr # 200-030-12
COMPARE
No cap included 33 mm 30
$102.55CAD / Each
COMPARE
Threaded cap 24 mm 7
$580.52CAD / Pkg of 10
COMPARE
Threaded cap 33 mm 15
$856.04CAD / Pkg of 10
COMPARE
Threaded cap 33 mm 22
$965.07CAD / Pkg of 10
COMPARE
Threaded cap 33 mm 30
$1,085.89CAD / Pkg of 10
Showing 1- 9 of 9