Sanitary Sampling Smart Gasket
Smart Gasket lets you take samples or readings from a sanitary line
  • Measure temperature, take a sample, or dose chemicals into your process line
  • Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
$83.25 - $185.65CAD / Each
46 variations of this product are available.
See All Product Options Below
46 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in)
Availability Pricing
Smart Gasket 3/4" Tri-Clamp, 1 Port, Viton, Each RK-30542-06 Mfr # G-TH-075-V-1
COMPARE
Viton 3/4
$87.67CAD / Each
Smart Gasket 1" Tri-Clamp, 1 Port, Silicone, Each RK-30542-08 Mfr # G-TH-100-S-1
COMPARE
Platinum-cured silicone 1
$124.50CAD / Each
Smart Gasket G-TH-100-E-1 1" Tri-Clamp, 1 Port, EPDM, Each RK-30542-09 Mfr # G-TH-100-E-1
COMPARE
EPDM 1
$118.61CAD / Each
Smart Gasket G-TH-100-V-1 1" Tri-Clamp, 1 Port, Viton, Each RK-30542-10 Mfr # G-TH-100-V-1
COMPARE
Viton 1
$113.45CAD / Each
Smart Gasket 1 1/2" Tri-Clamp, 1 Port, Silicone, Each RK-30542-12 Mfr # G-TH-150-S-1
COMPARE
Platinum-cured silicone 1-1/2
$111.24CAD / Each
Smart Gasket G-TH-200-E-1 2" Tri-Clamp, 1 Port, EPDM, Each RK-30542-17 Mfr # G-TH-200-E-1
COMPARE
EPDM 2
$125.24CAD / Each
Smart Gasket 1/2" Tri-Clamp, 2 Port, Silicone, Each RK-30543-00 Mfr # G-TH-050-S-2
COMPARE
Platinum-cured silicone 1/2
$102.40CAD / Each
Smart Gasket 3/4" Tri-Clamp, 2 Port, Viton, Each RK-30543-06 Mfr # G-TH-075-V-2
COMPARE
Viton 3/4
$97.24CAD / Each
Smart Gasket G-TH-100-E-2 1" Tri-Clamp, 2 Port, EPDM, Each RK-30543-09 Mfr # G-TH-100-E-2
COMPARE
EPDM 1
$123.03CAD / Each
Smart Gasket G-TH-150-S-2 1 1/2" Tri-Clamp, 2 Port, Silicone, Each RK-30543-12 Mfr # G-TH-150-S-2
COMPARE
Platinum-cured silicone 1-1/2
$138.50CAD / Each
Smart Gasket G-TH-200-E-2 2" Tri-Clamp, 2 Port, EPDM, Each RK-30543-17 Mfr # G-TH-200-E-2
COMPARE
EPDM 2
$142.18CAD / Each
Smart Gasket 1/2" Tri-Clamp, 3 Port, EPDM, Each RK-30544-01 Mfr # G-TH-050-E-3
COMPARE
EPDM 1/2
$113.45CAD / Each
Smart Gasket G-TH-075-S-3 3/4" Tri-Clamp, 3 Port, Silicone, Each RK-30544-04 Mfr # G-TH-075-S-3
COMPARE
Platinum-cured silicone 3/4
$111.98CAD / Each
Smart Gasket G-TH-075-E-3 3/4" Tri-Clamp, 3 Port, EPDM, Each RK-30544-05 Mfr # G-TH-075-E-3
COMPARE
EPDM 3/4
$113.45CAD / Each
Smart Gasket G-TH-150-E-3 1 1/2" Tri-Clamp, 3 Port, EPDM, Each RK-30544-13 Mfr # G-TH-150-E-3
COMPARE
EPDM 1-1/2
$156.18CAD / Each
Smart Gasket G-TH-150-V-3 1 1/2" Tri-Clamp, 3 Port, Viton, Each RK-30544-14 Mfr # G-TH-150-V-3
COMPARE
Viton 1-1/2
$142.18CAD / Each
Smart Gasket G-TH-200-S-3 2" Tri-Clamp, 3 Port, Silicone, Each RK-30544-16 Mfr # G-TH-200-S-3
COMPARE
Platinum-cured silicone 2
$148.81CAD / Each
Smart Gasket G-TH-150-S-4 1 1/2" Tri-Clamp, 4 Port, Silicone, Each RK-30545-12 Mfr # G-TH-150-S-4
COMPARE
Platinum-cured silicone 1-1/2
$184.17CAD / Each
Smart Gasket G-TH-150-E-4 1 1/2" Tri-Clamp, 4 Port, EPDM, Each RK-30545-13 Mfr # G-TH-150-E-4
COMPARE
EPDM 1-1/2
$185.65CAD / Each
Smart Gasket G-TH-150-V-4 1 1/2" Tri-Clamp, 4 Port, Viton, Each RK-30545-14 Mfr # G-TH-150-V-4
COMPARE
Viton 1-1/2
$165.02CAD / Each
Showing 1 - 20 of 46