SaniSure® Single-Use Sanitary Clamp Accessories
Fast, safe, secure, and cost-effective connections!
  • Even 360-degree compression to prevent leakage
  • Withstands pressures up to 100 psi (6.9 bar)
  • Reduces installation time
SaniSure clamps feature an innovative design for fast, safe, secure connections and offer a cost-effective solution for connecting single-use installations. Made of USP Class VI-approved nylon, these clamps are designed for single use applications but are durable enough to withstand repetitive use. Unlike traditional two-point load stainless steel sanitary clamps, the design of these clamps ensure even 360-degree compression over the entire fitting surface—no more leaks and no gasket intrusion in your process line! Their low-profile rounded edges reduce the risk of penetrating bag assemblies, and the evenly controlled compression allows easy connection for plastic-to plastic, plastic-to-stainless steel, or stainless steel-to-stainless steel coupling joints.
To use, simply insert the cable tie end in its mate a couple clicks; position clamp on ferrule; hold both clamp parts together; move clamp around the tri-clamp fitting joint and slide clamp closed. Complete the closure by using the clamping tool to bring it to the proper tension. The easy-to-use design reduces chances of risk associated with carpal tunnel or other wrist injuries and yields a tamper-proof connection that is not operator dependent. As a single-use device, they eliminate need for cleaning validations and reduce your overall downtime for installation.
$29.32 - $1,040.21CAD / Pkg of 50
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
SaniSure SS-CAB-TIE-750 Sanitary Clamps, Replacement 3/4 Sanitary Cable Tie, 50/Pk RK-30562-31 Mfr # SS-CAB-TIE-750
COMPARE
Sanitary Clamps, Replacement 3/4 Sanitary Cable Tie, 50/Pk
$57.83CAD / Pkg of 50
SaniSure SS-CAB-TIE-1500 Sanitary Clamps, Replacement 1-1/2 Sanitary Cable Tie, 50/Pk RK-30562-32 Mfr # SS-CAB-TIE-1500
COMPARE
Sanitary Clamps, Replacement 1-1/2 Sanitary Cable Tie, 50/Pk
$57.83CAD / Pkg of 50
SaniSure SS-MK-9 Pre-Set Clamp Installation Tool RK-30562-40 Mfr # SS-MK-9
COMPARE
Pre-Set Clamp Installation Tool
$1,040.21CAD / Each
SaniSure SS-CUT-170 Sanitary Clamp/Cable Tie Removal Tool RK-30562-42 Mfr # SS-CUT-170
COMPARE
Sanitary Clamp/Cable Tie Removal Tool
$29.32CAD / Each